Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-methoxy-2-(methylthio)pyrimidine 95%

Supplier: AMBEED, INC
Warning

A138455-1G A138455-100MG A138455-250MG A138455-1G
103802-616EA 44 USD InStock
103802-616 76972-468 76972-470 77315-646
4-Chloro-6-methoxy-2-(methylthio)pyrimidine 95%
4-Chloro-6-methoxy-2-(methylthio)pyrimidine
Precaución: for Research Use Only


Formula: C₆H₇ClN₂OS
Boiling Pt: 288 °C (760 mm Hg)
Melting Pt: 38…39 °C
Flash Pt: 128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10574952
CAS Number: 89466-42-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty