Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine 98%

Supplier: AMBEED, INC
Warning

A135115-1G A135115-100G A135115-10G A135115-250MG A135115-1G A135115-25G A135115-5G
103802-094EA 9 USD InStock
103802-094 77162-054 77162-056 77162-058 77314-880 77314-882 77314-884
4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine 98%
4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine
Precaución: for Research Use Only


Formula: C₆H₅ClN₄
Boiling Pt: 472.3 °C (760 mm Hg)
Melting Pt: 228…231 °C
Flash Pt: 239.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07369229
CAS Number: 84955-31-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty