Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-7-methoxyquinazoline 97%

Supplier: AMBEED, INC
Warning

A268967-1G A268967-5G A268967-10G A268967-1G A268967-5G A268967-250MG
103812-948EA 60 USD InStock
103812-948 103852-852 76816-152 77305-030 77305-032 77672-386
4-Chloro-7-methoxyquinazoline 97%
4-Chloro-7-methoxyquinazoline
Precaución: for Research Use Only


Formula: C₉H₇ClN₂O
MW: 194.618 g/mol
Boiling Pt: 323 °C (760 mm Hg)
Flash Pt: 149.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738758
CAS Number: 55496-52-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty