Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine 98%

Supplier: AMBEED, INC
Warning

A239055-5G A239055-10G A239055-25G
103811-260EA 9 USD InStock
103811-260 103851-056 76786-466 77304-132
4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine 98%
4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine
Precaución: for Research Use Only


Formula: C₁₃H₁₀ClN₃O₂S
Boiling Pt: 505.3 °C (760 mm Hg)
Flash Pt: 259.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09907939
CAS Number: 479633-63-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty