Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-8-fluoroquinazoline 95%

Supplier: AMBEED, INC
Warning

A144859-1G A144859-100MG A144859-250MG A144859-5G A144859-1G
103803-460EA 155 USD InStock
103803-460 76823-606 76823-608 76823-610 77316-792
4-Chloro-8-fluoroquinazoline 95%
4-Chloro-8-fluoroquinazoline
Precaución: for Research Use Only


Formula: C₈H₄ClFN₂
MW: 182.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09837193
CAS Number: 124429-27-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty