Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-8-methoxyquinazoline 95%

Supplier: AMBEED, INC
Warning

A111054-1G A111054-100MG A111054-10G A111054-250MG A111054-1G A111054-5G
103797-784EA 95 USD InStock
103797-784 103823-594 76781-086 76781-088 76781-090 77310-400 77310-402
4-Chloro-8-methoxyquinazoline 95%
4-Chloro-8-methoxyquinazoline
Precaución: for Research Use Only


Formula: C₉H₇ClN₂O
MW: 194.6200 g/mol
Boiling Pt: 323 °C (760 mm Hg)
Flash Pt: 149.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09954869
CAS Number: 154288-09-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty