Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-o-phenylenediamine 98%

Supplier: AMBEED, INC
Warning

A181336-5G A181336-25G A181336-5G A181336-100G A181336-10G A181336-500G
103845-968EA 9 USD InStock
103845-968 77083-492 77083-494 77287-222 77287-224 77287-226
4-Chloro-o-phenylenediamine 98%
4-Chloro-o-phenylenediamine
Precaución: for Research Use Only


Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Boiling Pt: 280
Melting Pt: 70…73 °C
Flash Pt: 135.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011691
CAS Number: 95-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty