Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorofuro[3,2-c]pyridine 97%

Supplier: AMBEED, INC
Warning

A101645-1G A101645-5G A101645-100MG A101645-10G A101645-250MG
103796-060EA 29 USD InStock
103796-060 103821-756 77282-780 77282-782 77282-784
4-Chlorofuro[3,2-c]pyridine 97%
4-Chlorofuro[3,2-c]pyridine
Precaución: for Research Use Only


Formula: C₇H₄ClNO
Boiling Pt: 242.8 °C (760 mm Hg)
Melting Pt: 41 °C
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179768
CAS Number: 31270-80-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty