Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloroisatin 97%

Supplier: AMBEED, INC
Warning

A164197-1G A164197-5G A164197-100G A164197-10G A164197-1G A164197-25G A164197-5G
103805-826EA 8 USD InStock
103805-826 103844-086 77286-540 77286-542 77320-234 77320-236 77320-238
4-Chloroisatin 97%
4-Chloroisatin
Precaución: for Research Use Only


Formula: C₈H₄ClNO₂
Melting Pt: 258 °C
Storage Temperature: Ambient
MDL Number: MFCD00129178
CAS Number: 6344-05-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty