Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloroisophthalic acid 95%

Supplier: AMBEED, INC
Warning

A224498-1G A224498-5G A224498-10G A224498-250MG A224498-1G A224498-5G
103819-112EA 20 USD InStock
103819-112 103850-004 77030-896 77030-898 77088-662 77088-664
4-Chloroisophthalic acid 95%
4-Chloroisophthalic acid
Precaución: for Research Use Only


Formula: C₈H₅ClO₄
MW: 200.58 g/mol
Boiling Pt: 424.9 °C (760 mm Hg)
Flash Pt: 210.8 °C
Storage Temperature: Ambient
MDL Number: MFCD12027303
CAS Number: 2845-85-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty