Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-N-methylaniline 97%

Supplier: AMBEED, INC
Danger

A122749-1G A122749-10G A122749-25G A122749-1G A122749-5G
103800-274EA 6 USD InStock
103800-274 77284-922 77284-924 77297-892 77312-388
4-Chloro-N-methylaniline 97%
4-Chloro-N-methylaniline
Precaución: for Research Use Only


Formula: C₇H₈ClN
MW: 141.6
Boiling Pt: 239
Density: 1.169
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000614
CAS Number: 932-96-7
UN: 2924
ADR: 3,8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty