Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloropicolinamide 97%

Supplier: AMBEED, INC
Warning

A214036-1G A214036-5G A214036-1G A214036-5G A214036-100G
103818-358EA 5 USD InStock
103818-358 103849-068 77087-566 77087-568 77152-536 77152-538
4-Chloropicolinamide 97%
4-Chloropicolinamide
Precaución: for Research Use Only


Formula: C₆H₅ClN₂O
Flash Pt: 134.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01085339
CAS Number: 99586-65-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty