Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloropyrimidine hydrochloride 96%

Supplier: AMBEED, INC
Warning

A274787-1G A274787-5G A274787-100G A274787-100MG A274787-10G A274787-250MG A274787-1G A274787-25G A274787-5G
103813-220EA 47 USD InStock
103813-220 103853-148 76787-256 76787-258 76787-260 76787-262 77305-174 77305-176 77305-178
4-Chloropyrimidine hydrochloride 96%
4-Chloropyrimidine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₃ClN₂·ClH
Melting Pt: 193…194 °C
Storage Temperature: Freezer
MDL Number: MFCD11870807
CAS Number: 179051-78-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty