Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorothiophene-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A141138-1G A141138-100MG A141138-250MG A141138-1G A141138-5G
103802-956EA 135 USD InStock
103802-956 77285-752 77285-754 77316-090 77316-092
4-Chlorothiophene-2-carboxylic acid 97%
4-Chlorothiophene-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₅H₃ClO₂S
Boiling Pt: 298 °C (760 mm Hg)
Flash Pt: 134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082876
CAS Number: 59614-95-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty