Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1H-pyrazole-4-carbonitrile 97%

Supplier: AMBEED, INC
Warning

A170085-1G A170085-5G A170085-250MG A170085-10G
103806-446EA 14 USD InStock
103806-446 103844-712 76973-966 77881-313
1-Methyl-1H-pyrazole-4-carbonitrile 97%
1-Methyl-1H-pyrazole-4-carbonitrile
Precaución: for Research Use Only


Formula: C₅H₅N₃
Boiling Pt: 230.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04969704
CAS Number: 66121-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty