Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-4-carbonitrile 97%

Supplier: AMBEED, INC
Warning

A208930-5G A208930-100G A208930-10G A208930-25G A208930-5G A208930-500G
103848-588EA 12 USD InStock
103848-588 76691-398 76691-400 77086-920 77086-922 77877-655
Indole-4-carbonitrile 97%
Indole-4-carbonitrile
Precaución: for Research Use Only


Formula: C₉H₆N₂
MW: 142.16
Boiling Pt: 350 °C (760 mm Hg)
Melting Pt: 117…121 °C
Flash Pt: 121.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00152045
CAS Number: 16136-52-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty