Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyclopropylphenol 98%

Supplier: AMBEED, INC
Danger

A157706-1G A157706-5G A157706-100MG A157706-250MG A157706-1G
103805-038EA 99 USD InStock
103805-038 103843-240 76783-694 76783-696 77319-078
4-Cyclopropylphenol 98%
4-Cyclopropylphenol
Precaución: for Research Use Only


Formula: C₉H₁₀O
Melting Pt: 57…58 °C
Storage Temperature: Ambient
MDL Number: MFCD06802405
CAS Number: 10292-61-2
UN: 2430
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty