Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A236314-5G A236314-25G A236314-5G A236314-100G A236314-10G A236314-1G
103850-850EA 20 USD InStock
103850-850 77089-898 77089-900 77193-232 77193-234 77193-236
4-Ethoxyphenylboronic acid 98%
4-Ethoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₁₁BO₃
Boiling Pt: 317.6 °C (760 mm Hg)
Melting Pt: 121…128 °C
Flash Pt: 145.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00674028
CAS Number: 22237-13-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty