Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethyl-1-naphthoic acid 95%

Supplier: AMBEED, INC
Warning

A197293-1G A197293-5G A197293-100G A197293-250MG A197293-25G A197293-1G A197293-5G
103816-908EA 15 USD InStock
103816-908 103847-530 76999-200 76999-202 76999-204 77085-526 77085-528
4-Ethyl-1-naphthoic acid 95%
4-Ethyl-1-naphthoic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₂O₂
MW: 200.23 g/mol
Boiling Pt: 379.3 °C (760 mm Hg)
Flash Pt: 173.2 °C
Storage Temperature: Ambient
MDL Number: MFCD17169910
CAS Number: 91902-58-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty