Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethynylpyridine hydrochloride 97%

Supplier: AMBEED, INC
Danger

A194057-1G A194057-5G A194057-100G A194057-1G A194057-25G A194057-5G
103816-614EA 13 USD InStock
103816-614 103847-224 77287-524 77302-382 77302-384 77302-386
4-Ethynylpyridine hydrochloride 97%
4-Ethynylpyridine hydrochloride


Formula: C₇H₆ClN
Storage Temperature: Refrigerator
MDL Number: MFCD03093912
CAS Number: 352530-29-1
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty