Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-1-naphthaldehyde 98%

Supplier: AMBEED, INC
Warning

A369157-1G A369157-5G A369157-100MG A369157-250MG A369157-1G
103826-446EA 31 USD InStock
103826-446 103837-460 76828-852 76828-854 76840-708
4-Fluoro-1-naphthaldehyde 98%
4-Fluoro-1-naphthaldehyde
Precaución: for Research Use Only


Formula: C₁₁H₇FO
Storage Temperature: Refrigerator
MDL Number: MFCD02261765
CAS Number: 172033-73-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty