Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-isopropoxyaniline 95%

Supplier: AMBEED, INC
Warning

A195801-1G A195801-5G A195801-1G A195801-250MG
103816-786EA 38 USD InStock
103816-786 103847-398 77085-344 77164-414 77164-416
4-Fluoro-2-isopropoxyaniline 95%
4-Fluoro-2-isopropoxyaniline
Precaución: for Research Use Only


Formula: C₉H₁₂FNO
MW: 169.196 g/mol
Boiling Pt: 237.0±20.0 °C (760 mm Hg)
Flash Pt: 97.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09738072
CAS Number: 148583-65-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty