Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-o-toluic acid 98%

Supplier: AMBEED, INC
Warning

A120160-1G A120160-100G A120160-10G A120160-25G A120160-500G A120160-1G A120160-5G
103799-894EA 5 USD InStock
103799-894 77271-444 77271-446 77271-448 77271-450 77297-742 77311-998
4-Fluoro-o-toluic acid 98%
4-Fluoro-o-toluic acid
Precaución: for Research Use Only


Formula: C₈H₇FO₂
MW: 154.14
Boiling Pt: 265.6 °C (760 mm Hg)
Melting Pt: 168…172 °C
Flash Pt: 114.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04115880
CAS Number: 321-21-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty