Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-methylphenol 98%

Supplier: AMBEED, INC
Warning

A238706-5G A238706-100G A238706-10G A238706-500G A238706-25G A238706-5G
103851-032EA 7 USD InStock
103851-032 76771-352 76771-354 76771-356 77090-164 77090-166
4-Fluoro-2-methylphenol 98%
4-Fluoro-2-methylphenol
Precaución: for Research Use Only


Formula: C₇H₇FO
Boiling Pt: 198 °C (760 mm Hg)
Melting Pt: 37…38 °C
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00075088
CAS Number: 452-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty