Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-methylbenzaldehyde 97% (by GC)

Supplier: AMBEED, INC
Warning

A161691-1G A161691-5G A161691-100G A161691-10G A161691-1G A161691-25G A161691-5G
103805-518EA 9 USD InStock
103805-518 103843-758 76689-850 76689-852 77300-548 77319-748 77319-750
4-Fluoro-3-methylbenzaldehyde 97% (by GC)
4-Fluoro-3-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₇FO
Boiling Pt: 203.6 °C (760 mm Hg)
Flash Pt: 79.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01631431
CAS Number: 135427-08-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty