Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Fluoro-3'-nitroacetophenone 98%

Supplier: AMBEED, INC
Warning

A164972-5G A164972-100G A164972-10G A164972-500G A164972-25G A164972-5G
103844-182EA 10 USD InStock
103844-182 76689-986 76689-988 76689-990 77320-378 77320-380
4'-Fluoro-3'-nitroacetophenone 98%
4'-Fluoro-3'-nitroacetophenone
Precaución: for Research Use Only


Formula: C₈H₆FNO₃
MW: 183.14
Boiling Pt: 261.2 °C (760 mm Hg)
Melting Pt: 46…50 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00115369
CAS Number: 400-93-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty