Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-nitrophenylboronic acid 95%

Supplier: AMBEED, INC
Warning

A108032-1G A108032-5G A108032-10G A108032-250MG A108032-1G A108032-5G
103797-282EA 35 USD InStock
103797-282 103823-056 77270-678 77270-680 77309-896 77309-898
4-Fluoro-3-nitrophenylboronic acid 95%
4-Fluoro-3-nitrophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₅BFNO₄
Boiling Pt: 367 °C (760 mm Hg)
Flash Pt: 175.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08277236
CAS Number: 352530-22-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty