Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluorophthalic anhydride 97%

Supplier: AMBEED, INC
Warning

A190235-1G A190235-5G A190235-100G A190235-10G A190235-500G A190235-1G A190235-25G A190235-5G
103816-272EA 5 USD InStock
103816-272 103846-874 76769-858 76769-860 76769-862 77084-662 77084-664 77084-666
3-Fluorophthalic anhydride 97%
3-Fluorophthalic anhydride
Precaución: for Research Use Only


Formula: C₈H₃FO₃
MW: 166.11 g/mol
Boiling Pt: 303.3 °C (760 mm Hg)
Melting Pt: 159…163 °C
Flash Pt: 132.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00039696
CAS Number: 652-39-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty