Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluororesorcinol 98%

Supplier: AMBEED, INC
Warning

A244561-1G A244561-5G A244561-1G A244561-10G A244561-250MG A244561-5G
103811-586EA 16 USD InStock
103811-586 103851-418 77090-628 77288-808 77288-810 77304-322
4-Fluororesorcinol 98%
4-Fluororesorcinol
Precaución: for Research Use Only


Formula: C₆H₅FO₂
MW: 128.10 g/mol
Melting Pt: 96…100 °C
Flash Pt: 99.4 °C
Storage Temperature: Freezer
MDL Number: MFCD03789074
CAS Number: 103068-41-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty