Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Formylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A340775-25G A340775-5G A340775-100G A340775-10G A340775-500G
103836-792EA 9 USD InStock
103836-792 76839-806 76839-808 77142-090 77142-092 77142-094
4-Formylphenylboronic acid 98%
4-Formylphenylboronic acid
Precaución: For research use only.


Formula: C₇H₇BO₃
Melting Pt: 237…242 °C
Flash Pt: 164.013 °C
Storage Temperature: Ambient
MDL Number: MFCD00151823
CAS Number: 87199-17-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty