Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydrazinobenzoic acid 98%

Supplier: AMBEED, INC
Warning

A221774-5G A221774-10G A221774-500G A221774-25G A221774-5G A221774-100G
103849-822EA 5 USD InStock
103849-822 76770-848 76770-850 77088-420 77088-422 77877-689
4-Hydrazinobenzoic acid 98%
4-Hydrazinobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₈N₂O₂
Boiling Pt: 377.2 °C (760 mm Hg)
Melting Pt: 218 °C
Flash Pt: 181.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00007581
CAS Number: 619-67-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty