Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3,5-dimethylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A100539-1G A100539-5G A100539-1G A100539-5G
103795-500EA 6 USD InStock
103795-500 103821-190 77296-742 77296-744
4-Hydroxy-3,5-dimethylbenzoic acid 98%
4-Hydroxy-3,5-dimethylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
Boiling Pt: 331.3 °C (760 mm Hg)
Melting Pt: 221…226 °C
Flash Pt: 168.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00016536
CAS Number: 4919-37-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty