Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3-iodobenzonitrile 97%

Supplier: AMBEED, INC
Warning

A656593-5G A656593-1G A656593-100MG A656593-250MG
103840-420EA 319 USD InStock
103840-420 76844-608 77158-026 77158-028
4-Hydroxy-3-iodobenzonitrile 97%
4-Hydroxy-3-iodobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₄INO
MW: 245.017 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11054826
CAS Number: 2296-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty