Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-4-(trifluoromethyl)cyclohexanecarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A197499-1G A197499-100MG A197499-250MG
103816-926EA 182 USD InStock
103816-926 76975-104 76975-106
4-Hydroxy-4-(trifluoromethyl)cyclohexanecarboxylic acid 95%
4-Hydroxy-4-(trifluoromethyl)cyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₁F₃O₃
MW: 212.1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13659409
CAS Number: 1163729-47-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty