Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dihydroxy-6-methylpyridine 97%

Supplier: AMBEED, INC
Warning

A675987-100G A675987-10G A675987-25G A675987-1G A675987-5G
103840-522EA 9 USD InStock
103840-522 76819-910 76819-912 76819-914 76844-722 76844-724
2,4-Dihydroxy-6-methylpyridine 97%
2,4-Dihydroxy-6-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₇NO₂
MW: 125.13 g/mol
Melting Pt: 324…327 °C
Storage Temperature: Ambient
MDL Number: MFCD01860849
CAS Number: 3749-51-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty