Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzamidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A150074-1G A150074-5G A150074-250MG A150074-1G A150074-25G A150074-5G A150074-10G
103804-116EA 6 USD InStock
103804-116 103842-250 77190-770 77299-772 77299-774 77299-776 77746-044
4-Hydroxybenzamidine hydrochloride 95%
4-Hydroxybenzamidine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₉ClN₂O
Boiling Pt: 313.9 °C (760 mm Hg)
Melting Pt: 223…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00136405
CAS Number: 38148-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty