Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A337115-25G A337115-1000G A337115-100G A337115-100MG A337115-500G
103836-712EA 5 USD InStock
103836-712 76839-714 77114-974 77114-976 77114-978 77114-980
4-Hydroxybenzyl alcohol 98%
4-Hydroxybenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₈O₂
Boiling Pt: 252 °C (760 mm Hg)
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004658
CAS Number: 623-05-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty