Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Aminomethyl)phenol 97%

Supplier: AMBEED, INC
Warning

A690871-25G A690871-5G A690871-100G
103840-580EA 10 USD InStock
103840-580 76844-796 76844-798 77293-202
4-(Aminomethyl)phenol 97%
4-(Aminomethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₉NO
MW: 123.16 g/mol
Boiling Pt: 262.4 °C (760 mm Hg)
Melting Pt: 116…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19690971
CAS Number: 696-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty