Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxyisoquinolin-1(2H)-one 95%

Supplier: AMBEED, INC
Warning

A172738-1G A172738-100MG A172738-250MG A172738-1G
103806-742EA 157 USD InStock
103806-742 76824-558 76824-560 77082-248
4-Hydroxyisoquinolin-1(2H)-one 95%
4-Hydroxyisoquinolin-1(2H)-one
Precaución: for Research Use Only


Formula: C₉H₇NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01646237
CAS Number: 30081-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty