Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxyphthalic acid 98%

Supplier: AMBEED, INC
Warning

A271679-1G A271679-5G A271679-25G A271679-5G A271679-100G A271679-10G A271679-500G A271679-1G
103813-058EA 9 USD InStock
103813-058 103852-970 77092-750 77092-752 77238-750 77238-752 77238-754 77305-092
4-Hydroxyphthalic acid 98%
4-Hydroxyphthalic acid


Formula: C₈H₆O₅
Boiling Pt: 468.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00013984
CAS Number: 610-35-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty