Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxyphthalonitrile 95%

Supplier: AMBEED, INC
Warning

A127906-1G A127906-100G A127906-10G A127906-1G A127906-25G A127906-5G
103801-036EA 10 USD InStock
103801-036 76766-980 76766-982 77313-308 77313-310 77313-312
4-Hydroxyphthalonitrile 95%
4-Hydroxyphthalonitrile
Precaución: for Research Use Only


Formula: C₈H₄N₂O
Boiling Pt: 394.6 °C (760 mm Hg)
Melting Pt: 218 °C
Flash Pt: 192.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00191653
CAS Number: 30757-50-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty