Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Quinolinol 97%

Supplier: AMBEED, INC
Warning

A207237-5G A207237-5G A207237-100G A207237-10G A207237-500G A207237-25G
103848-454EA 9 USD InStock
103848-454 77086-722 77260-970 77260-972 77260-974 77303-024
4-Quinolinol 97%
4-Quinolinol
Precaución: for Research Use Only


Formula: C₉H₇NO
Boiling Pt: 261.5 °C (760 mm Hg)
Melting Pt: 200…202 °C
Flash Pt: 122.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00006777
CAS Number: 611-36-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty