Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-2-quinolinecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A141971-1G A141971-100MG A141971-250MG A141971-25MG A141971-25G A141971-5G
103803-084EA 32 USD InStock
103803-084 76798-346 76798-348 76798-350 77316-286 77624-176
4-Hydroxy-2-quinolinecarboxylic acid 97%
4-Hydroxy-2-quinolinecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₇NO₃
Boiling Pt: 358.4 °C (760 mm Hg)
Melting Pt: 275 °C
Storage Temperature: Ambient
MDL Number: MFCD00149476
CAS Number: 492-27-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty