Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodo-o-toluidine 98%

Supplier: AMBEED, INC
Warning

A157846-5G A157846-100G A157846-1G A157846-5G A157846-10G
103843-256EA 8 USD InStock
103843-256 77190-978 77190-980 77319-090 77746-080
4-Iodo-o-toluidine 98%
4-Iodo-o-toluidine
Precaución: Keep in a dark place, sealed in dry, room temperature


Formula: C₇H₈IN
MW: 233.05 g/mol
Boiling Pt: 279
Melting Pt: 86…89 °C
Flash Pt: 122.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00025299
CAS Number: 13194-68-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty