Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-iodobenzene-1,2-diamine 97%

Supplier: AMBEED, INC
Warning

A200389-1G A200389-5G A200389-100MG A200389-250MG A200389-1G
103817-198EA 13 USD InStock
103817-198 103847-826 77192-262 77192-264 77302-680
4-iodobenzene-1,2-diamine 97%
4-iodobenzene-1,2-diamine
Precaución: for Research Use Only


Formula: C₆H₇IN₂
Melting Pt: 73…77 °C
Flash Pt: 158.8 °C
Storage Temperature: Freezer
MDL Number: MFCD08669466
CAS Number: 21304-38-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty