Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodotetrahydropyran 95%

Supplier: AMBEED, INC
Warning

A155400-1G A155400-5G A155400-100G A155400-10G A155400-250MG A155400-1G A155400-25G A155400-5G
103804-758EA 19 USD InStock
103804-758 103842-954 77162-936 77162-938 77162-940 77318-670 77318-672 77318-674
4-Iodotetrahydropyran 95%
4-Iodotetrahydropyran
Precaución: Keep in a dark place, sealed in dry, at 2…8°C


Formula: C₅H₉IO
Boiling Pt: 208.1 °C (760 mm Hg)
Flash Pt: 79.689 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797467
CAS Number: 25637-18-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty