Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-1H-pyrazole 95%

Supplier: AMBEED, INC
Warning

A114591-1G A114591-100MG A114591-250MG A114591-25G A114591-1G A114591-5G
103798-428EA 58 USD InStock
103798-428 103824-268 77094-190 77094-192 77094-194 77094-196 77311-084 77311-086
4-Methoxy-1H-pyrazole 95%
4-Methoxy-1H-pyrazole
Precaución: for Research Use Only


Formula: C₄H₆N₂O
MW: 98.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22394063
CAS Number: 14884-01-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty