Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-1,5-dihydro-2H-pyrrol-2-one 95%

Supplier: AMBEED, INC
Warning

A197206-1G A197206-5G A197206-100MG A197206-250MG A197206-1G A197206-25G A197206-5G
103816-900EA 12 USD InStock
103816-900 103847-520 77012-536 77012-538 77085-504 77085-506 77085-508
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one 95%
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one
Precaución: for Research Use Only


Formula: C₅H₇NO₂
Melting Pt: 129…132 °C
Flash Pt: 165.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00071564
CAS Number: 69778-83-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty