Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-2,4-dioxobutanoic acid 96%

Supplier: AMBEED, INC
Warning

A199231-1G A199231-1G A199231-100MG A199231-250MG
103817-084EA 192 USD InStock
103817-084 103847-708 77085-750 77164-530 77164-532
4-Methoxy-2,4-dioxobutanoic acid 96%
4-Methoxy-2,4-dioxobutanoic acid
Precaución: for Research Use Only


Formula: C₅H₆O₅
Boiling Pt: 246.8±40.0 °C (760 mm Hg)
Flash Pt: 103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19229811
CAS Number: 13192-05-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty