Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-o-toluic acid 98%

Supplier: AMBEED, INC
Warning

A109169-1G A109169-100G A109169-10G A109169-250MG A109169-25G A109169-500G A109169-1G A109169-5G
103797-460EA 6 USD InStock
103797-460 103823-252 77283-824 77283-826 77283-828 77283-830 77283-832 77310-070 77310-072
4-Methoxy-o-toluic acid 98%
4-Methoxy-o-toluic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
Boiling Pt: 294.3 °C (760 mm Hg)
Melting Pt: 177…181 °C
Flash Pt: 118.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00020291
CAS Number: 6245-57-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty